Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-11
Entrez Gene ID
180938
Gene Name
protein-coding
Gene Aliases
CELE_K11G12.1
Chromosome Location
Chr.X: 6713577 - 6716381 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029618.5 NP_001024789.1 1 - 2 283 89
NM_001029619.7 NP_001024790.1 1 - 2 282 89

View Full Product Details