Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
odr-10
Entrez Gene ID
180959
Gene Name
Serpentine receptor class r-10
Gene Aliases
CELE_C53B7.5
Chromosome Location
Chr.X: 6864313 - 6865939 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076756.4 NP_509157.1 6 - 7 760 95
GenBank mRNA U49449.1 - 6 - 7 765 95

View Full Product Details