Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-6
Entrez Gene ID
180961
Gene Name
Netrin unc-6
Gene Aliases
CELE_F41C6.1
Chromosome Location
Chr.X: 6889674 - 6897147 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076764.5 NP_509165.1 2 - 3 280 61

View Full Product Details