Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tyra-2
Entrez Gene ID
180970
Gene Name
protein-coding
Gene Aliases
CELE_F01E11.5
Chromosome Location
Chr.X: 6996853 - 7005575 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029350.7 NP_001024521.1 9 - 10 1296 92
NM_001029351.6 NP_001024522.1 9 - 10 1078 92
NM_001038448.5 NP_001033537.1 9 - 10 1086 92

View Full Product Details