Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-7
Entrez Gene ID
180983
Gene Name
DNaJ domain (prokaryotic heat shock protein)
Gene Aliases
CELE_C55B6.2
Chromosome Location
Chr.X: 7194564 - 7198299 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076808.6 NP_509209.1 8 - 9 1331 72

View Full Product Details