Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-14
Entrez Gene ID
181012
Gene Name
Homeobox protein ceh-14
Gene Aliases
CELE_F46C8.5
Chromosome Location
Chr.X: 7527769 - 7532732 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076872.4 NP_509273.1 7 - 8 675 67
GenBank mRNA AF244368.1 - 7 - 8 689 67

View Full Product Details