Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sto-1
- Entrez Gene ID
-
181017
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F08C6.4
- Chromosome Location
-
Chr.X: 7562824 - 7564814 on Build WBcel235
- Gene Symbol
- F08C6.5
- Entrez Gene ID
-
184175
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F08C6.5
- Chromosome Location
-
Chr.X: 7559405 - 7564812 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076879.5 |
NP_509280.2 |
6 - 7 |
2433 |
66 |
View Full Product Details