Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sul-3
Entrez Gene ID
183778
Gene Name
Sulfatase domain-containing protein
Gene Aliases
CELE_C54D2.4
Chromosome Location
Chr.X: 7827085 - 7834217 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373376.2 NP_001360719.1 9 - 9 1448 121

View Full Product Details