Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-11
Entrez Gene ID
191845
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_R09F10.6
Chromosome Location
Chr.X: 8317637 - 8319050 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077004.1 NP_509405.1 3 - 4 320 169

View Full Product Details