Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sec-15
Entrez Gene ID
181131
Gene Name
Exocyst complex component 6
Gene Aliases
CELE_C28G1.3
Chromosome Location
Chr.X: 8831737 - 8835715 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077095.10 NP_509496.3 10 - 11 2227 81

View Full Product Details