Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ifp-1
Entrez Gene ID
181182
Gene Name
Intermediate filament protein ifp-1
Gene Aliases
CELE_C43C3.1
Chromosome Location
Chr.X: 9675824 - 9680169 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077227.7 NP_509628.1 8 - 9 2161 78

View Full Product Details