Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpy-22
Entrez Gene ID
181194
Gene Name
Mediator of RNA polymerase II transcription subunit 12
Gene Aliases
CELE_F47A4.2
Chromosome Location
Chr.X: 9810202 - 9822273 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077244.4 NP_509645.1 5 - 6 883 115

View Full Product Details