Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-37
Entrez Gene ID
181208
Gene Name
protein-coding
Gene Aliases
CELE_C17G1.6
Chromosome Location
Chr.X: 9944844 - 9949068 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029242.6 NP_001024413.1 4 - 5 596 79
NM_001029243.6 NP_001024414.1 4 - 5 595 79
GenBank mRNA AY912076.1 - 4 - 5 610 79

View Full Product Details