Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-12
Entrez Gene ID
181263
Gene Name
protein-coding
Gene Aliases
CELE_F11A1.3
Chromosome Location
Chr.X: 10644440 - 10666875 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029376.6 NP_001024547.1 5 - 6 450 141
NM_001047774.5 NP_001041239.1 4 - 5 348 141
NM_001381088.1 NP_001366986.1 3 - 4 273 141
GenBank mRNA AF136238.1 - 5 - 6 483 141
AF136239.1 - 3 - 4 280 141

View Full Product Details