The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129669.4 | NP_001123141.2 | 28 - 29 | 4237 | 113 |
NM_001373489.2 | NP_001360637.1 | 28 - 29 | 4216 | 113 | |
NM_001373491.2 | NP_001360639.1 | 28 - 29 | 4246 | 113 | |
NM_001373492.2 | NP_001360635.1 | 28 - 29 | 4267 | 113 | |
NM_001373493.2 | NP_001360636.1 | 29 - 30 | 4387 | 113 | |
NM_001373494.2 | NP_001359720.1 | 29 - 30 | 4408 | 113 | |
NM_001373495.2 | NP_001360638.1 | 29 - 30 | 4417 | 113 |