Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
plc-1
Entrez Gene ID
181274
Gene Name
protein-coding
Gene Aliases
CELE_F31B12.1
Chromosome Location
Chr.X: 10794857 - 10838650 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129669.4 NP_001123141.2 29 - 30 4444 93
NM_001373489.2 NP_001360637.1 29 - 30 4423 93
NM_001373491.2 NP_001360639.1 29 - 30 4453 93
NM_001373492.2 NP_001360635.1 29 - 30 4474 93
NM_001373493.2 NP_001360636.1 30 - 31 4594 93
NM_001373494.2 NP_001359720.1 30 - 31 4615 93
NM_001373495.2 NP_001360638.1 30 - 31 4624 93

View Full Product Details