Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
plc-1
Entrez Gene ID
181274
Gene Name
protein-coding
Gene Aliases
CELE_F31B12.1
Chromosome Location
Chr.X: 10794857 - 10838650 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129669.4 NP_001123141.2 10 - 11 963 60
NM_001373489.2 NP_001360637.1 10 - 11 963 60
NM_001373491.2 NP_001360639.1 10 - 11 963 60
NM_001373492.2 NP_001360635.1 10 - 11 963 60
NM_001373493.2 NP_001360636.1 10 - 11 963 60
NM_001373494.2 NP_001359720.1 10 - 11 963 60
NM_001373495.2 NP_001360638.1 10 - 11 963 60

View Full Product Details