Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pgp-5
Entrez Gene ID
181276
Gene Name
protein-coding
Gene Aliases
CELE_C05A9.1
Chromosome Location
Chr.X: 10854216 - 10859297 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270187.3 NP_001257116.1 17 - 18 3464 88
NM_001270188.3 NP_001257117.1 14 - 15 3121 88

View Full Product Details