Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pgp-8
Entrez Gene ID
181277
Gene Name
P-GlycoProtein related
Gene Aliases
CELE_T21E8.3
Chromosome Location
Chr.X: 10859301 - 10864422 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077410.6 NP_509811.2 2 - 3 256 109

View Full Product Details