Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-15
Entrez Gene ID
181291
Gene Name
protein-coding
Gene Aliases
CELE_F58A3.2
Chromosome Location
Chr.X: 11016400 - 11023421 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313426.3 NP_001300355.1 10 - 11 1956 64
NM_001313427.4 NP_001300356.1 10 - 11 1956 64
NM_001313435.5 NP_001300364.1 11 - 11 1792 64
NM_001373496.2 NP_001360406.1 10 - 11 1955 64
NM_001381095.1 NP_001368580.1 10 - 11 1759 64
NM_001381096.1 NP_001368579.1 10 - 11 1759 64
NM_001381098.1 NP_001367004.1 10 - 11 1759 64
NM_001383640.1 10 - 11 1956 64
GenBank mRNA U39761.1 - 10 - 11 1924 64

View Full Product Details