Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alr-1
Entrez Gene ID
181302
Gene Name
Homeobox domain-containing protein
Gene Aliases
CELE_R08B4.2
Chromosome Location
Chr.X: 11121871 - 11125620 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077459.6 NP_509860.1 2 - 3 313 71

View Full Product Details