Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hst-2
Entrez Gene ID
181309
Gene Name
Heparan sulfate 2-O-sulfotransferase hst-2
Gene Aliases
CELE_C34F6.4
Chromosome Location
Chr.X: 11204103 - 11207155 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077470.6 NP_509871.2 6 - 7 815 146
GenBank mRNA AY552606.1 - 6 - 7 748 146

View Full Product Details