Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vhl-1
Entrez Gene ID
181321
Gene Name
von Hippel-Lindau tumor suppressor homolog
Gene Aliases
CELE_F08G12.4
Chromosome Location
Chr.X: 11307356 - 11308518 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077488.6 NP_509889.1 1 - 2 151 61

View Full Product Details