Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-14
Entrez Gene ID
181337
Gene Name
protein-coding
Gene Aliases
CELE_T25C12.1
Chromosome Location
Chr.X: 11465557 - 11485871 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077515.4 NP_509916.2 10 - 11 1483 113
NM_077516.6 NP_509917.1 8 - 9 1576 113

View Full Product Details