Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frm-3
Entrez Gene ID
186110
Gene Name
protein-coding
Gene Aliases
CELE_H05G16.1
Chromosome Location
Chr.X: 11559405 - 11585126 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001351892.4 NP_001338849.1 8 - 8 939 127
NM_001375142.3 NP_001361874.1 7 - 8 909 127

View Full Product Details