Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lsd-1
Entrez Gene ID
181369
Gene Name
Probable lysine-specific histone demethylase 1
Gene Aliases
CELE_T08D10.2
Chromosome Location
Chr.X: 11947427 - 11951937 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077599.6 NP_510000.2 5 - 6 1010 69

View Full Product Details