Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-13
Entrez Gene ID
187971
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_T01H10.1
Chromosome Location
Chr.X: 12109135 - 12111300 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077622.4 NP_510023.2 5 - 6 707 154

View Full Product Details