Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
otpl-8
Entrez Gene ID
186752
Gene Name
OToPetrin-Like
Gene Aliases
CELE_H19J13.1
Chromosome Location
Chr.X: 12266953 - 12270941 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077644.6 NP_510045.2 3 - 4 296 74

View Full Product Details