Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-46
Entrez Gene ID
184606
Gene Name
Serpentine receptor class delta-46
Gene Aliases
CELE_F17A2.10
Chromosome Location
Chr.X: 12332095 - 12333302 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077664.1 NP_510065.1 4 - 5 701 73

View Full Product Details