Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nekl-3
Entrez Gene ID
181398
Gene Name
Serine/threonine-protein kinase nekl-3
Gene Aliases
CELE_F19H6.1
Chromosome Location
Chr.X: 12389531 - 12391480 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077679.8 NP_510080.2 7 - 8 844 65
GenBank mRNA DQ645955.1 - 7 - 8 860 65

View Full Product Details