Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myo-2
Entrez Gene ID
181404
Gene Name
Myosin-2
Gene Aliases
CELE_T18D3.4
Chromosome Location
Chr.X: 12468061 - 12475348 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077691.5 NP_510092.2 4 - 5 897 74

View Full Product Details