Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc-13
Entrez Gene ID
181406
Gene Name
Rab-GAP TBC domain-containing protein
Gene Aliases
CELE_F45E6.3
Chromosome Location
Chr.X: 12485053 - 12490970 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373530.1 NP_001359553.1 4 - 5 760 94

View Full Product Details