Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-19
Entrez Gene ID
181407
Gene Name
SKN-1 Dependent Zygotic transcript
Gene Aliases
CELE_F45E6.6
Chromosome Location
Chr.X: 12502710 - 12503338 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077696.6 NP_510097.2 1 - 2 71 66

View Full Product Details