Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pgp-13
Entrez Gene ID
184845
Gene Name
P-GlycoProtein related
Gene Aliases
CELE_F22E10.2
Chromosome Location
Chr.X: 12738754 - 12744111 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373542.2 NP_001359545.1 1 - 2 297 86

View Full Product Details