Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-25
Entrez Gene ID
181432
Gene Name
protein-coding
Gene Aliases
CELE_F11C1.6
Chromosome Location
Chr.X: 13008487 - 13013965 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029379.4 NP_001024550.1 5 - 6 554 120
NM_001029380.7 NP_001024551.1 5 - 6 540 120

View Full Product Details