Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
asp-4
Entrez Gene ID
181444
Gene Name
Aspartic protease 4
Gene Aliases
CELE_R12H7.2
Chromosome Location
Chr.X: 13218970 - 13221462 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077790.7 NP_510191.1 3 - 4 431 120

View Full Product Details