Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hpl-1
Entrez Gene ID
181450
Gene Name
Chromo domain-containing protein
Gene Aliases
CELE_K08H2.6
Chromosome Location
Chr.X: 13245390 - 13247044 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077798.5 NP_510199.1 2 - 3 214 88
GenBank mRNA AF056580.1 - 2 - 3 198 88

View Full Product Details