Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-43
Entrez Gene ID
187577
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_R04D3.9
Chromosome Location
Chr.X: 13303905 - 13305844 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322642.1 NP_001309448.1 5 - 6 771 120

View Full Product Details