Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sad-1
Entrez Gene ID
181471
Gene Name
protein-coding
Gene Aliases
CELE_F15A2.6
Chromosome Location
Chr.X: 13487546 - 13500668 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083291.4 NP_001076760.1 8 - 9 1536 70
NM_001083292.4 NP_001076761.1 8 - 9 1542 70
GenBank mRNA AF316542.1 - 8 - 9 1524 70

View Full Product Details