Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-36
Entrez Gene ID
186981
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_K04C1.6
Chromosome Location
Chr.X: 14229668 - 14234466 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077969.2 NP_510370.1 2 - 3 262 89

View Full Product Details