Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ram-5
Entrez Gene ID
181550
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_T24C2.1
Chromosome Location
Chr.X: 14550138 - 14556297 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078021.4 NP_510422.2 6 - 7 619 97
GenBank mRNA AF218866.1 - 6 - 7 641 97

View Full Product Details