Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-31A3
Entrez Gene ID
189467
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_Y17G9B.3
Chromosome Location
Chr.IV: 4761049 - 4762885 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068236.4 NP_500637.2 1 - 2 109 106

View Full Product Details