Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-48
Entrez Gene ID
180188
Gene Name
C-type LECtin
Gene Aliases
CELE_C14A6.1
Chromosome Location
Chr.V: 18157878 - 18159614 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075146.2 NP_507547.1 2 - 3 636 132

View Full Product Details