Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-64
Entrez Gene ID
191982
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T28A11.1
Chromosome Location
Chr.V: 3283257 - 3284930 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071506.2 NP_503907.1 2 - 2 822 102

View Full Product Details