Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gsp-4
Entrez Gene ID
171960
Gene Name
Serine/threonine-protein phosphatase PP1-delta
Gene Aliases
CELE_T03F1.5
Chromosome Location
Chr.I: 3838237 - 3839653 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001384033.1 2 - 2 352 117

View Full Product Details