Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-2
Entrez Gene ID
177872
Gene Name
Cuticle collagen 2
Gene Aliases
CELE_W01B6.7
Chromosome Location
Chr.IV: 10084439 - 10085451 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069428.4 NP_501829.1 1 - 1 181 151
GenBank mRNA AY347575.1 - 1 - 1 168 151

View Full Product Details