Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-42
Entrez Gene ID
3565714
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F14F8.11
Chromosome Location
Chr.V: 16692125 - 16693832 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028628.1 NP_001023799.1 1 - 2 326 141

View Full Product Details