Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-87
Entrez Gene ID
3565609
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F38B7.8
Chromosome Location
Chr.V: 11564873 - 11566771 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028726.1 NP_001023897.1 1 - 2 70 156

View Full Product Details