Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-1
Entrez Gene ID
3564925
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_T11F9.18
Chromosome Location
Chr.V: 11485858 - 11487266 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028964.4 NP_001024135.2 1 - 2 169 145

View Full Product Details