Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnc-10
Entrez Gene ID
3565147
Gene Name
CaeNaCin (Caenorhabditis bacteriocin)
Gene Aliases
CELE_VK10D6R.1
Chromosome Location
Chr.V: 11183224 - 11183643 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029007.2 NP_001024178.1 1 - 2 76 63

View Full Product Details